1-Aminocyclopentanecarboxylic acid 98%

Supplier: Ambeed
Warning

A224871-10G A224871-1G A224871-25G A224871-5G A224871-100G
77165-322EA 19 USD
77165-322 77165-324 77165-326 77165-328 77612-184
1-Aminocyclopentanecarboxylic acid 98%
1-Aminocyclopentanecarboxylic acid

Caution: for Research Use Only

Formula: C₆H₁₁NO₂
MDL Number: MFCD00001381
CAS Number: 52-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR