1,10-Phenanthroline hydrochloride monohydrate 99%
Supplier: Ambeed
|
Danger
|
Synonyms:
1,10-Phenanthroline Chloride Monohydrate, 1,10-Phenanthrolinium chloride monohydrate, 1,10-Phenanthroline monohydrochloride monohydrate
A218543-10G
A218543-25G
77192-832EA
11
USD
77192-832
77192-834
77192-836
1,10-Phenanthroline hydrochloride monohydrate 99%
1,10-Phenanthroline hydrochloride monohydrate
Caution: for Research Use Only
Formula:
C₁₂H₉ClN₂·1H₂O MW: 234.68 g/mol |
MDL Number:
MFCD00150061 CAS Number: 18851-33-7 UN: 2811 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...