L(+)-Glutamic acid 98%

Supplier: Ambeed
Warning

Synonyms: L-Glutamic acid(S)-(+)-1-Aminopropane-1,3-dicarboxylic acidL(+)-α-Glutamic acidH-Glu-OH(S)-(+)-Glutamic acid(S)-(+)-α-Glutamic acidL(+)-1-Aminopropane-1,3-dicarboxylic acid(S)-(+)-2-Aminopentanedioic acidL(+)-2-Aminopentanedioic acid

A211917-25G A211917-1000G A211917-500G A211917-100G
77164-968EA 7.21 USD
77164-968 77164-966 77164-970 77164-972
L(+)-Glutamic acid 98%
L(+)-Glutamic acid

Caution: for Research Use Only

Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
MDL Number: MFCD00002634
CAS Number: 56-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR