N-Methyl-D-aspartic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Me-D-Asp-OH
, N-Methyl-D-asparaginic Acid
, (R)-2-(Methylamino)succinic acid
A188283-50MG
A188283-1MG
A188283-100MG
77164-146EA
14
USD
77164-146
77164-142
77164-144
N-Methyl-D-aspartic acid 98%
N-Methyl-D-aspartic acid
Caution: for Research Use Only
Formula:
C₅H₉NO₄ MW: 147.13 g/mol |
MDL Number:
MFCD00004226 CAS Number: 6384-92-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...