5-Bromo-2-fluoroisonicotinic acid 95%

Supplier: Ambeed
Warning

A186957-1G A186957-250MG A186957-5G
77164-094EA 43 USD
77164-094 77164-096 77164-098
5-Bromo-2-fluoroisonicotinic acid 95%
5-Bromo-2-fluoroisonicotinic acid

Caution: for Research Use Only

Formula: C₆H₃BrFNO₂
MDL Number: MFCD13185803
CAS Number: 1214377-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR