2-Amino-5-thiazolecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Aminothiazole-5-carboxylic acid

A164763-10G A164763-1G A164763-5G A164763-25G
77163-322EA 29 USD
77163-322 77163-324 77163-326 77892-114
2-Amino-5-thiazolecarboxylic acid 95%
2-Amino-5-thiazolecarboxylic acid

Caution: for Research Use Only

Formula: C₄H₄N₂O₂S
MDL Number: MFCD06203554
CAS Number: 40283-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR