10-Acetylphenothiazine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(10H-Phenothiazin-10-yl)ethanone, 10-Acetyl-10H-phenothiazine

A155426-100MG A155426-250MG A155426-1G
77162-942EA 7 USD
77162-942 77162-944 77892-067
10-Acetylphenothiazine 95%
10-Acetylphenothiazine

Caution: for Research Use Only

Formula: C₁₄H₁₁NOS
MW: 241.31 g/mol
MDL Number: MFCD00022264
CAS Number: 1628-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR