Bergenin 98%

Supplier: Ambeed
Warning

A123838-10G A123838-10MG A123838-1G A123838-1MG A123838-250MG A123838-25MG A123838-50MG A123838-5G A123838-5MG
77189-770EA 915.99 USD
77189-770 77189-772 77189-774 77189-776 77189-778 77189-780 77189-782 77189-784 77189-786
Bergenin 98%
Bergenin

Caution: for Research Use Only

Formula: C₁₄H₁₆O₉
MDL Number: MFCD00133120
CAS Number: 477-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR