Undec-10-enoyl chloride 99%

Supplier: Ambeed
Danger

A122363-25G A122363-5G
77189-720EA 37 USD
77189-720 77189-722
Undec-10-enoyl chloride 99%
Undec-10-enoyl chloride

Caution: for Research Use Only

Formula: C₁₁H₁₉ClO
MDL Number: MFCD00000772
CAS Number: 38460-95-6
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR