Ninhydrin 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2-Dihydroxyindan-1,3-dion
, Trioxohydrindene monohydrate
, Ninhydrin monohydrate
, 2,2-Dihydroxy-1,3-indanedione
, Indanetrione hydrate
, 2,2-Dihydroxyindane-1,3-dione
, 1,2,3-Indantrione monohydrate
A109112-500G
A109112-10G
A109112-1G
A109112-100G
A109112-25G
A109112-5G
77188-968EA
11.33
USD
77188-968
77188-966
77188-976
77188-974
77188-972
77188-970
Ninhydrin 98%
Ninhydrin
Caution: for Research Use Only
Formula:
C₉H₆O₄ MW: 178.14 g/mol |
MDL Number:
MFCD00003791 CAS Number: 485-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...