Nile red 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
A819053-1G
A819053-250MG
A819053-5G
77159-336EA
150
USD
77159-336
77159-338
77159-340
Nile red 98%
Nile red
Caution: for Research Use Only
Formula:
C₂₀H₁₈N₂O₂ |
MDL Number:
MFCD00011639 CAS Number: 7385-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...