Sodium dihydrogen phosphate monohydrate 99%

Supplier: Ambeed
Warning

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

A809930-1000G A809930-100G A809930-25G A809930-500G
77159-274EA 12 USD
77159-274 77159-276 77159-278 77159-280
Sodium dihydrogen phosphate monohydrate 99%
Sodium dihydrogen phosphate monohydrate

Caution: for Research Use Only

Formula: NaH₂PO₄·H₂O
MDL Number: MFCD00149208
CAS Number: 10049-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR