p-Anisaldehyde 99%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxybenzaldehyde

A796070-1000G A796070-100G A796070-25G A796070-500G
77159-200EA 37 USD
77159-200 77159-202 77159-204 77159-206
p-Anisaldehyde 99%
p-Anisaldehyde

Caution: for Research Use Only

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 140
Flash Pt: 110
MDL Number: MFCD00003385
CAS Number: 123-11-5
Merck Index: 13,00666

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR