Manganese (II) acetylacetonate 98%

Supplier: Ambeed
Danger

Synonyms: Bis(2,4-pentanedionato-O,O')manganese (II), Manganese (II) bis(2,4-pentanedionato-O,O'), Manganese(II) bis(acetylacetonate), Manganese (II) 2,4-pentanedionate

A698624-25G A698624-5G
77158-416EA 11 USD
77158-416 77158-418
Manganese (II) acetylacetonate 98%
Manganese (II) acetylacetonate

Caution: for Research Use Only

Formula: C₁₀H₁₄MnO₄
MW: 253.16 g/mol
MDL Number: MFCD00000022
CAS Number: 14024-58-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR