D-(+)-Galactose 95%

Supplier: Ambeed
Warning

A665119-100G A665119-25G A665119-500G A665119-1000G
77158-096EA 17 USD
77158-096 77158-098 77158-100 77686-633
D-(+)-Galactose 95%
D(+)-Galactose

Caution: for Research Use Only

Formula: C₆H₁₂O₆
MDL Number: MFCD00151230
CAS Number: 59-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR