Manganese(II) sulfate monohydrate 98%

Supplier: Ambeed
Danger

A642501-1000G A642501-500G A642501-100G
77157-966EA 6.18 USD
77157-966 77613-992 77613-994 77686-590
Manganese(II) sulfate monohydrate 98%
Manganese (II) sulphate monohydrate

Caution: for Research Use Only

Formula: MnSO₄·1H₂O
MDL Number: MFCD00149159
CAS Number: 10034-96-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR