N-Phthalyl-L-tryptophan 98%

Supplier: Ambeed
Warning

A548158-10MG A548158-1MG A548158-50MG A548158-5MG A548158-100MG A548158-1G A548158-250MG
77157-132EA 40 USD
77157-132 77157-134 77157-136 77157-138 77686-408 77686-409 77686-410
N-Phthalyl-L-tryptophan 98%
N-Phthalyl-L-tryptophan

Caution: for Research Use Only

Formula: C₁₉H₁₄N₂O₄
MDL Number: MFCD08705332
CAS Number: 48208-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR