2-(4-Iodophenyl)-5,5-dimethyl-1,3,2-dioxaborinane ≥95%

Supplier: Ambeed
Warning

A520144-250MG A520144-5G
77156-864EA 11 USD
77156-864 77156-866
2-(4-Iodophenyl)-5,5-dimethyl-1,3,2-dioxaborinane ≥95%
2-(4-Iodophenyl)-5,5-dimethyl-1,3,2-dioxaborinane

Caution: for Research Use Only

Formula: C₁₁H₁₄BIO₂
MDL Number: MFCD26394871
CAS Number: 5572-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR