2,3-Dichloro-6-(trifluoromethyl)benzyl alcohol 95+%

Supplier: Ambeed
Warning

Synonyms: (2,3-Dichloro-6-(trifluoromethyl)phenyl)methanol

A463393-1G A463393-250MG A463393-5G
77156-230EA 55 USD
77156-230 77156-232 77156-234
2,3-Dichloro-6-(trifluoromethyl)benzyl alcohol 95+%
2,3-Dichloro-6-(trifluoromethyl)benzyl alcohol

Caution: for Research Use Only

Formula: C₈H₅Cl₂F₃O
MDL Number: MFCD06660274
CAS Number: 886501-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR