n-Hexadecane 98%

Supplier: Ambeed
Danger

Synonyms: Hexadecane, Cetane

A426120-25G A426120-100G
77155-866EA 7 USD
77155-866 77613-318
n-Hexadecane 98%
n-Hexadecane

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₄CH₃
MW: 226.45 g/mol
Boiling Pt: 287
Melting Pt: 18
Flash Pt: 135
MDL Number: MFCD00008998
CAS Number: 544-76-3
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR