BIX 01294 98%

Supplier: Ambeed
Warning

A337768-10MG A337768-1MG A337768-50MG A337768-5MG A337768-100MG A337768-1G A337768-250MG
77154-850EA 9 USD
77154-850 77154-852 77154-854 77154-856 77685-966 77685-967 77685-968
BIX 01294 98%
BIX 01294

Caution: for Research Use Only

Formula: C₂₈H₃₈N₆O₂
MDL Number: MFCD14560563
CAS Number: 935693-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR