Lenvatinib (E7080) 98%

Supplier: Ambeed
Warning

Synonyms: 4-[3-chloro-4-(cyclopropylcarbamoylamino)phenoxy]-7-methoxyquinoline-6-carboxamide, Lenvatinib, 4-(3-Chloro-4-(3-cyclopropylureido)phenoxy)-7-methoxyquinoline-6-carboxamide

A336250-100MG A336250-1G A336250-5G A336250-250MG A336250-5MG A336250-10G A336250-10MG
77154-796EA 11 USD
77154-796 77154-798 77154-800 77612-928 77612-930 77685-962 77685-963
Lenvatinib (E7080) 98%
Lenvatinib (E7080)

Caution: for Research Use Only

Formula: C₂₁H₁₉ClN₄O₄
MW: 426.85 g/mol
MDL Number: MFCD16038644
CAS Number: 417716-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR