Eosin Y 97% BS

Supplier: Ambeed
Warning

Synonyms: Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-

A308289-100G A308289-25G A308289-500G A308289-5G
77154-308EA 66 USD
77154-308 77154-310 77154-312 77154-314
Eosin Y 97% BS
Eosin Y

Caution: for Research Use Only

Formula: C₂₀H₈Br₄O₅
MW: 647.89 g/mol
Melting Pt: 295
MDL Number: MFCD00036189
CAS Number: 15086-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR