Lithium sulfate 99.9%
Supplier: Ambeed
|
Warning
|
A297016-25G
A297016-500G
A297016-100G
A297016-1000G
77154-134EA
14
USD
77154-134
77154-136
77685-846
77900-583
Lithium sulfate 99.9%
Lithium sulphate
Caution: for Research Use Only
Formula:
Li₂O₄S MW: 109.95 g/mol Boiling Pt: 1377 Melting Pt: 859 |
MDL Number:
MFCD00011086 CAS Number: 10377-48-7 Merck Index: 12,05569 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...