RG7422 98+%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(4-((2-(2-aminopyrimidin-5-yl)-7-methyl-4-morpholinothieno[3,2-d]pyrimidin-6-yl)methyl)piperazin-1-yl)-2-hydroxypropan-1-one, Apitolisib, GDC-0980

A281340-1MG A281340-50MG A281340-5MG
77153-836EA 44 USD
77153-836 77153-838 77153-840
RG7422 98+%
RG7422

Caution: for Research Use Only

Formula: C₂₃H₃₀N₈O₃S
MW: 498.6 g/mol
MDL Number: MFCD20274520
CAS Number: 1032754-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR