1-Methylpiperazine 98%

Supplier: Ambeed
Danger

A199162-25G A199162-100G
77152-090EA 14 USD
77152-090 77892-269
1-Methylpiperazine 98%
1-Methylpiperazine

Caution: for Research Use Only

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 138
Melting Pt: -6
Flash Pt: 34
MDL Number: MFCD00005966
CAS Number: 109-01-3
UN: 2924
ADR: 3,8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR