AZD-8186 98+%

Supplier: Ambeed
Warning

Synonyms: AZD8186, azd-8186, (R)-8-(1-((3,5-difluorophenyl)amino)ethyl)-N,N-dimethyl-2-morpholino-4-oxo-4H-chromene-6-carboxamide

A160272-10MG A160272-1MG A160272-25MG A160272-50MG A160272-5MG
77150-840EA 13 USD
77150-840 77150-842 77150-844 77150-846 77150-848
AZD-8186 98+%
AZD-8186

Caution: for Research Use Only

Formula: C₂₄H₂₅F₂N₃O₄
MW: 457.47 g/mol
MDL Number: MFCD27987908
CAS Number: 1627494-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR