L(-)-Proline 95%
Supplier: Ambeed
|
Warning
|
A110254-1000G
A110254-100G
A110254-1MG
A110254-25G
A110254-500G
77148-668EA
43
USD
77148-668
77148-670
77148-672
77148-674
77148-676
L(-)-Proline 95%
L(-)-Proline
Caution: for Research Use Only
Formula:
C₅H₉NO₂ |
MDL Number:
MFCD00064318 CAS Number: 147-85-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...