Control Slides, Normal Human Tissue, Salivary Gland
Supplier: Mortech Manufacturing
SC914-100 SC914-25
76948-094CS 1281.94 USD
76948-094 76948-096
Control Slides, Normal Human Tissue, Salivary Gland
Slides Microscopy Control Slides
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using formalin fixed paraffin embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properlyMore Product Information