Lighthouse Electric Fog/Mist Unit, 15 Gallon
Supplier: Unimed
Lighthouse
F15G516130
77007-822EA 3741.97 USD
77007-822
Lighthouse Electric Fog/Mist Unit, 15 Gallon
Spray Guns
This 120 VAC electric unit lets users apply disinfectants and other chemistry with a handheld wand. With a built-in air compressor, simply plug the unit into a standard wall outlet and get to work.
  • Draws from pre-mixed solution
  • Powered by electricity (120 VAC)
  • Built-in air compressor
  • Quick-change pump
  • Hinged lid
  • Front-facing drain
  • In-line strainer protects pump from debris
Ideal for fogging/misting in areas without compressed air. Unit features a handheld wand with 25' (7.6 m) of coiled discharge hose and built-in air compressor. A see-through tank provides a 15 gallon (56.8 L) capacity. Hinged back plate provides tool-free access to components.
Color coding options include 7 lid colors (black, blue, green, yellow, red, purple, and orange).