HydroFoamer HydroSprayer Device
Supplier: Unimed
HydroFoamer HydroSprayer
HPGF412483
77007-796EA 182.57 USD
77007-796
HydroFoamer HydroSprayer Device
Spray Guns
Increase labor productivity with the HydroFoamer and HydroSprayer. These easy-to-use spraying and foaming devices can be utilized anywhere a hose can reach.
  • Proven, reliable components
  • Non-electric
  • No installation required
  • Compact, easy to store
  • Enhanced safety due to reduced exposure to chemicals
  • Maximum cleaning product performance
Chemical concentrates will perform at their peak because they are mixed only as they are applied. Just fill the concentrate container and connect the dispenser to a water source. Quick application, properly diluted chemicals, and minimal waste make the HydroFoamer and HydroSprayer devices some of the most popular products in the industry.