Control Slides, Special Stains, Melanin
Supplier: Mortech Manufacturing
SC821-10 SC821-100 SC821-20
76947-940CS 230.66 USD
76947-940 76947-942 76947-944
Control Slides, Special Stains, Melanin
Slides Microscopy Control Slides
All positive control slides are on Mopec IHC enhanced hydrophilic adhesive slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 microns
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt whenMore Product Information