(-)-N,N'-(1R,2R)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide) 98%

Supplier: Ambeed
Warning

Synonyms: N,N'-(trans-Cyclohexane-1,2-diyl)dipicolinamide, (R,R)-DACH-Pyridyl Trost Ligand

A892230-1G A892230-250MG A892230-5G
77147-062EA 43.25 USD
77147-062 77147-064 77147-066
(-)-N,N'-(1R,2R)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide) 98%
(-)-N,N'-(1R,2R)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide)

Caution: for Research Use Only

Formula: C₁₈H₂₀N₄O₂
MDL Number: MFCD02684543
CAS Number: 218290-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR