6-Bromo-2-fluoro-3-(trifluoromethoxy)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
A868656-1G
A868656-5G
77146-978EA
39.14
USD
77146-978
77146-980
6-Bromo-2-fluoro-3-(trifluoromethoxy)benzoic acid 97%
6-Bromo-2-fluoro-3-(trifluoromethoxy)benzoic acid
Caution: for Research Use Only
Formula:
C₈H₃BrF₄O₃ |
MDL Number:
MFCD28134151 CAS Number: 524674-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...