(S)-(-)-1-Phenylethyl isocyanate 98%

Supplier: Ambeed
Danger

Synonyms: (S)-1-phenylethylisocyanate(S)-(-)-α-Phenethyl isocyanate(S)-(-)-α-Methylbenzyl isocyanate(S)-(-)-1-Phenethyl isocyanate

A841433-5G
77146-868EA 28.84 USD
77146-868
(S)-(-)-1-Phenylethyl isocyanate 98%
(S)-(-)-1-Phenylethyl isocyanate

Caution: for Research Use Only

Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00002036
CAS Number: 14649-03-7
UN: 2206
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR