Diphenylboric acid-2-aminoethyl ester (2-APB) 98%

Supplier: Ambeed
Warning

Synonyms: 2-Aminoethyl diphenylborinate, 2-((Diphenylboryl)oxy)ethanamine

A682989-10G A682989-1G A682989-1MG A682989-25G A682989-5G A682989-100G
77145-566EA 124.63 USD
77145-566 77145-568 77145-570 77145-572 77145-574 77617-844
Diphenylboric acid-2-aminoethyl ester (2-APB) 98%
Diphenylboric acid 2-aminoethyl ester (2-APB)

Caution: for Research Use Only

Formula: C₁₄H₁₆BNO
MDL Number: MFCD00014823
CAS Number: 524-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR