(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate, (-)-1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate, (1Z,5Z)-cycloocta-1,5-diene, (2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane, rhodium, trifluoromethanesulfonate, (R,R)-Et-DUPHOS-Rh, 1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, (-)-1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, (1Z,5Z)-cycloocta-1,5-diene, (2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane, rhodium, trifluoromethanesulphonate

A663961-100MG A663961-250MG A663961-50MG
77145-396EA 181.27 USD
77145-396 77145-398 77145-400
(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate 98%
(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate

Caution: for Research Use Only

Formula: C₃₁H₄₈F₃O₃P₂RhS⁻
MW: 722.63 g/mol
MDL Number: MFCD00269860
CAS Number: 136705-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR