1-Allyl-3-methylimidazolium bromide 98%

Supplier: Ambeed
Warning

A592094-100G A592094-1G A592094-5G
77144-812EA 59.75 USD
77144-812 77144-814 77144-816
1-Allyl-3-methylimidazolium bromide 98%
1-Allyl-3-methylimidazolium bromide

Caution: for Research Use Only

Formula: C₇H₁₁N₂Br
MW: 203.08 g/mol
MDL Number: MFCD08277003
CAS Number: 31410-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR