Vinylene carbonate 98% stabilised

Supplier: Ambeed
Danger

Synonyms: 1,3-Dioxol-2-one

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A527298-10G A527298-25G
77144-224EA 6.18 USD
77144-224 77144-226
Vinylene carbonate 98% stabilised
Vinylene carbonate

Caution: for Research Use Only

Formula: C₃H₂O₃
MDL Number: MFCD00005380
CAS Number: 872-36-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR