2-Amino-4-chlorobenzothiazole 97%

Supplier: Ambeed
Warning

A523294-10G A523294-25G A523294-5G
77144-182EA 21.63 USD
77144-182 77144-184 77144-186
2-Amino-4-chlorobenzothiazole 97%
2-Amino-4-chlorobenzothiazole

Caution: for Research Use Only

Formula: C₇H₅ClN₂S
MW: 184.65 g/mol
MDL Number: MFCD00005791
CAS Number: 19952-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR