5-Acetylthiophene-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Acetylthiophene-2-carboxylic Acid, 5-Acetyl-2-thenoic acid
A469662-100G
A469662-1G
A469662-25G
A469662-5G
77143-648EA
239.99
USD
77143-648
77143-650
77143-652
77143-654
5-Acetylthiophene-2-carboxylic acid 98%
5-Acetylthiophene-2-carboxylic acid
Caution: for Research Use Only
Formula:
C₇H₆O₃S MW: 170.19 g/mol |
MDL Number:
MFCD00055512 CAS Number: 4066-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...