AT-13387 ≥98%

Supplier: Ambeed
Warning

Synonyms: (2,4-Dihydroxy-5-isopropylphenyl)(5-((4-methylpiperazin-1-yl)methyl)isoindolin-2-yl)methanone, Onalespib, AT13387, (2,4-dihydroxy-5-propan-2-ylphenyl)-[5-[(4-methylpiperazin-1-yl)methyl]-1,3-dihydroisoindol-2-yl]methanone, (2,4-dihydroxy-5-isopropylphenyl)(5-((4-methylpiperazin-1-yl)methyl)isoindolin-2-yl)methanone

A389427-10MG A389427-5MG A389427-100MG A389427-50MG
77142-700EA 94.76 USD
77142-700 77142-702 77686-079 77686-080
AT-13387 ≥98%
AT-13387

Caution: for Research Use Only

Formula: C₂₄H₃₁N₃O₃
MDL Number: MFCD18633198
CAS Number: 912999-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR