2-Acetamido-6-hydroxypurine 98%

Supplier: Ambeed
Warning

Synonyms: N-2-Acetylguanine, N-Acetylguanine

A285763-10G A285763-1G A285763-5G A285763-100G
77141-244EA 60.78 USD
77141-244 77141-246 77141-248 77550-756
2-Acetamido-6-hydroxypurine 98%
2-Acetamido-6-hydroxypurine

Caution: for Research Use Only

Formula: C₇H₇N₅O₂
MDL Number: MFCD00078201
CAS Number: 19962-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR