2-Oxo-1-pyrrolidineacetic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Oxo-1-pyrrolidinyl)acetic acid
A232501-100MG
A232501-1G
A232501-250MG
A232501-5G
77140-100EA
14.42
USD
77140-100
77140-102
77140-104
77140-106
2-Oxo-1-pyrrolidineacetic acid 97%
2-Oxo-1-pyrrolidineacetic acid
Caution: for Research Use Only
Formula:
C₆H₉NO₃ MW: 143.14 g/mol |
MDL Number:
MFCD00962836 CAS Number: 53934-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...