Amino-PEG2-acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Amino-PEG2-C2-acid, α-amine-ω-propionic acid diethylene glycol, H2N-PEG2-CH2CH2COOH, 3-(2-(2-Aminoethoxy)ethoxy)propanoic acid, 3-[2-(2-Aminoethoxy)ethoxy]-propanoic acid
A185794-100MG
A185794-250MG
A185794-1G
A185794-5G
A185794-25G
77138-892EA
11.34
USD
77138-892
77138-894
77550-190
77567-708
77685-435
Amino-PEG2-acid 95%
Amino-PEG2-acid
Caution: For research use only.
Formula:
C₇H₁₅NO₄ MW: 177.198 g/mol |
MDL Number:
MFCD20926377 CAS Number: 791028-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...