6-Fluoronaphthalen-1-ol 95%

Supplier: Ambeed
Warning

A184712-100MG A184712-1G
77138-846EA 65.93 USD
77138-846 77567-694
6-Fluoronaphthalen-1-ol 95%
6-Fluoronaphthalen-1-ol

Caution: For research use only.

Formula: C₁₀H₇FO
MW: 162.16 g/mol
MDL Number: MFCD15528810
CAS Number: 804498-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR