DL-Norvaline 98%

Supplier: Ambeed
Warning

A148781-100G A148781-10G A148781-25G A148781-500G
77137-686EA 31.94 USD
77137-686 77137-688 77137-690 77137-692
DL-Norvaline 98%
DL-Norvaline

Caution: for Research Use Only

Formula: C₅H₁₁NO₂
MDL Number: MFCD00064420
CAS Number: 760-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR