A-1210477 98%

Supplier: Ambeed
Warning

Synonyms: 7-(5-((4-(4-(N,N-dimethylsulphamoyl)piperazin-1-yl)phenoxy)methyl)-1,3-dimethyl-1H-pyrazol-4-yl)-1-(2-morpholinoethyl)-3-(3-(naphthalen-1-yloxy)propyl)-1H-indole-2-carboxylic acid, 7-(5-((4-(4-(N,N-dimethylsulfamoyl)piperazin-1-yl)phenoxy)methyl)-1,3-dimethyl-1H-pyrazol-4-yl)-1-(2-morpholinoethyl)-3-(3-(naphthalen-1-yloxy)propyl)-1H-indole-2-carboxylic acid

A116740-10MG A116740-1MG A116740-25MG A116740-50MG A116740-5MG A116740-100MG
77136-300EA 57.67 USD
77136-300 77136-302 77136-304 77136-306 77136-308 77685-032
A-1210477 98%
A-1210477

Caution: for Research Use Only

Formula: C₄₆H₅₅N₇O₇S
MW: 850.04 g/mol
MDL Number: MFCD28399118
CAS Number: 1668553-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR