(-)-10-Camphorsulfonic acid 98%
Supplier: Ambeed
|
Danger
|
A108406-25G
A108406-100G
77135-828EA
14.42
USD
77135-828
77614-826
(-)-10-Camphorsulfonic acid 98%
L(-)-Camphorsulfonic acid
Caution: for Research Use Only
Formula:
C₁₀H₁₆O₄S |
MDL Number:
MFCD00064158 CAS Number: 35963-20-3 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...