3,5-Difluoro-2-methoxyphenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (3,5-Difluoro-2-methoxyphenyl)boronic acid, 3,5-Difluoro-2-methoxybenzeneboronic acid

A105846-250MG A105846-5G A105846-1G
77135-692EA 18.04 USD
77135-692 77135-694 77684-959
3,5-Difluoro-2-methoxyphenylboronic acid 96%
3,5-Difluoro-2-methoxyphenylboronic acid

Caution: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.94 g/mol
MDL Number: MFCD03095353
CAS Number: 737000-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR